Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBCE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555341 Supplier Invitrogen™ Supplier No. PA555341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERGKHDGSHE GTVYFKCRHP TGGSFIRPNK VNFGTDFLTA IKNRYVLEDG PEEDRKEQIV TIGNKPVETI GFDSIMKQQS QLSKLQEVSL RNCAVSCAGE Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.

Cofactor E is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBCE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBCE
Gene Accession No. Q15813
Gene Alias 2610206D02Rik; beta-tubulin cofactor E; C530005D02Rik; HRD; hypoparathyroidism, growth and mental retardation, and dysmorphism; hypothetical protein LOC664760; KCS; KCS1; pac2; pmn; Tbce; Tbce protein; tubulin folding cofactor E; tubulin-folding cofactor E; Tubulin-specific chaperone E; tubulin-specific chaperone E-like protein; Unknown (protein for MGC:133938); zgc:123075
Gene Symbols TBCE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBCE. Recombinant protein control fragment (Product #RP-94104).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6905
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.