Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBCE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110461 Supplier Invitrogen™ Supplier No. PA5110461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEKLPSLRAL SCLRNPLTKE DKEAETARLL IIASIGQLKT LNKCEILPEE RRRAELDYRK AFGNEWKQAG GHKDPEKNRL SEEFLTAHPR YQFLCLKYGA PEDWELKTQQ PLMLKNQLLT LKIKYPHQLD QKVLEKQLPG SMTIQKVKGL LSRLLKVPVS DLLLSYESPK KPGREIELEN DLKSLQFYSV ENGDCLLVRW.

Cofactor E is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBCE
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TBCE
Gene Accession No. Q15813
Gene Alias 2610206D02Rik; beta-tubulin cofactor E; C530005D02Rik; HRD; hypoparathyroidism, growth and mental retardation, and dysmorphism; hypothetical protein LOC664760; KCS; KCS1; pac2; pmn; Tbce; Tbce protein; tubulin folding cofactor E; tubulin-folding cofactor E; Tubulin-specific chaperone E; tubulin-specific chaperone E-like protein; Unknown (protein for MGC:133938); zgc:123075
Gene Symbols TBCE
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 328-527 of human TBCE (NP_003184.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6905
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.