Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBCEL Monoclonal Antibody (OTI2A9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502672S


Description
TBCEL Monoclonal antibody specifically detects TBCEL in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
TBCEL | |
Monoclonal | |
0.84 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O96004 | |
Mouse | |
Affinity chromatography | |
RUO | |
9421 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2A9 | |
Unconjugated | |
HAND1 | |
catastrophin; D330014K23; E130107N23Rik; El; E-like; leucine rich repeat containing 35; leucine-rich repeat-containing protein 35; Lrrc35; RGD1308880; Tbcel; tubulin folding cofactor E like; tubulin folding cofactor E-like; tubulin-specific chaperone cofactor E-like protein; tubulin-specific chaperone e-like | |
Full length human recombinant protein of human HAND1 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction