Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBCEL Monoclonal Antibody (OTI3H3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525481
Description
TBCEL Monoclonal Antibody for Western Blot, Flow
Acts as a regulator of tubulin stability.
Specifications
TBCEL | |
Monoclonal | |
0.45 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5QJ74 | |
TBCEL | |
Full length human recombinant protein of TBCEL produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI3H3 | |
Unconjugated | |
TBCEL | |
catastrophin; D330014K23; E130107N23Rik; El; E-like; leucine rich repeat containing 35; leucine-rich repeat-containing protein 35; Lrrc35; RGD1308880; Tbcel; tubulin folding cofactor E like; tubulin folding cofactor E-like; tubulin-specific chaperone cofactor E-like protein; tubulin-specific chaperone e-like | |
Mouse | |
Affinity Chromatography | |
RUO | |
219899 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction