Learn More
Promega TBK1 Kinase Enzyme System, 1mg
Catalog No. PRV3992 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3992 1 mg
1 options
Catalog No. PRV3992 Supplier Promega Supplier No. V3992
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human TBK1 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. TBK1, also known as NAK or NFkB-activating kinase, is an upstream protein kinase that can phosphorylate and activate the IkB kinases.
  • Easily Screen and Profile TBK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C