Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBK1, p Ser172 Antibody (SR2251), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321592100UL
Description
TBK1 Monoclonal antibody specifically detects TBK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
TBK1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation | |
SR2251 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1 | |
A synthesized peptide derived from human phospho-TBK1 [Ser172] (Uniprot #: Q9UHD2) | |
100 μg | |
Phospho Specific, Protein Kinase, Signal Transduction | |
29110 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction