Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519968
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519968 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519968 Supplier Invitrogen™ Supplier No. PA519968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~84kDa. A suggested positive control is MOLT4 cell lysate. PA5-19968 can be used with blocking peptide PEP-0093.

TBK1 serine/threonine kinase is a member of the IKK family and is involved in NF-kappa B activation and IRF3 signaling. TBK1 is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-KB mediates the expression of a great variety of genes in response to extracellular stimuli. NF-KB is associated with IKB proteins in the cell cytoplasm, which inhibit NF-KB activity. Phosphorylation of I-KB by IKB kinase (IKK) complex leads to degradation of I-KB and activation of NF-KB. The IKK complex contains IKKa, IKK beta, and IKKK. A novel IKK related kinase was recently identified and designated TBK1 (TANK-binding kinase 1), NAK (NF-KB-activating kinase), and T2K. NAK/TBK1 activates IKK beta through direct phosphorylation. NAK/TBK1 is activated by growth factors and PMA and mediates IKK and NF-kB activation in response to growth factors. NAK/TBK1 functions upstream of NIK and the IKK complex. NAK/TBK1 is also critical in protecting embryonic liver from apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBK1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TBK1
Gene Accession No. Q9UHD2, Q9WUN2
Gene Alias 1200008B05Rik; AI462036; AW048562; FLJ11330; FTDALS4; NAK; NF-kappa-B-activating kinase; NF-kB-activating kinase; serine/threonine protein kinase TBK1; serine/threonine-protein kinase TBK1; T2K; TANK binding kinase 1; TANK-binding kinase 1; tbk; TBK1
Gene Symbols TBK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids form near the carboxy terminus of human NAK/TBK1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29110, 56480
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.