Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TBK1 PROTAC™ 3i
GREENER_CHOICESDP

Catalog No. 72595 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-595 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 72-595 Supplier Tocris Bioscience™ Supplier No. 7259/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent TANK-binding kinase 1 (TBK1) PROTAC™

TBK1 PROTAC® 3i is a potent TANK-binding kinase 1 (TBK1) PROTAC™ Degrader (DC50 = 12 nM, Dmax = 96%). Exhibits >50-fold selectivity for TBK1 over the closely related IKKε. Comprises a ligand for von-Hippel Lindau (VHL) protein joined by a linker to a TBK1-targeting moiety. Brings about near complete degradation of TBK1 in mutant K-Ras and wild-type cancer cell lines with no significant effect of proliferation.

Negative control TBK1 control PROTAC® 4 (Cat. No. 7260) also available.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

Specifications

Chemical Name or Material (2 S,4 R)-1-(-18-(4-((5-Bromo-4-((3-(N-methylcyclobutanecarboxamido)propyl)amino)pyrimidin-2-yl)amino)phenoxy)-2-(tert-butyl)-4-oxo-6,10,15-trioxa-3-azaoctadecanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
CAS 2052306-13-3
Quantity 5 mg
Target Active Degraders
Molecular Formula C53H74BrN9O9S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.