Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBK1 Rabbit anti-Human, Mouse, Rat, Clone: 4G6C7, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $494.50
Specifications
Antigen | TBK1 |
---|---|
Clone | 4G6C7 |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TBK1 Monoclonal antibody specifically detects TBK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
TBK1 | |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Protein Kinase, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
29110 | |
IgG | |
Affinity purified |
4G6C7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TBK1 (Q9UHD2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title