Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBKBP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TBKBP1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TBKBP1 Polyclonal specifically detects TBKBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TBKBP1 | |
Polyclonal | |
Rabbit | |
Human | |
A7MCY6 | |
9755 | |
This antibody was developed against a recombinant protein corresponding to amino acids: ELKQLQETRAQDLASNQSERDMAWVKRVGDDQVNLALAYTELTEELGRLRELSSLQGRILRTLLQEQARSG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ProSAPiP2, SINTBAD, TBKBP1 TBK1 binding protein 1 | |
TBKBP1 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title