Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBL1XR1 Monoclonal Antibody (OTI2A8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811698

Description
The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation.Specifications
TBL1XR1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8BHJ5, Q9BZK7 | |
Tbl1xr1 | |
Full length human recombit protein of human TBL1XR1 produced in E.coli. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
OTI2A8 | |
Unconjugated | |
TBL1XR1 | |
C21, DC42, IRA1, MRD41, TBLR1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
365755, 79718, 81004 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction