Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBL2 Mouse anti-Human, Clone: 1D5C2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TBL2 |
---|---|
Clone | 1D5C2 |
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6739
![]() ![]() |
Proteintech Group Inc
663071IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a member of the beta-transducin protein family. Most proteins of the beta-transducin family are involved in regulatory functions. This protein is possibly involved in some intracellular signaling pathway. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.Specifications
TBL2 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TBL2, transducin (beta) like 2, Transducin beta like protein 2, WBSCR13, WS betaTRP | |
TBL2 | |
IgA | |
Affinity Chromatography | |
Antibody |
1D5C2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9Y4P3 | |
26608 | |
TBL2 Fusion Protein Ag24225 | |
Primary | |
-20°C | |
TBL2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title