Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599726 Supplier Invitrogen™ Supplier No. PA599726
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TBL2.

This gene encodes a member of the beta-transducin protein family. Most proteins of the beta-transducin family are involved in regulatory functions. This protein is possibly involved in some intracellular signaling pathway. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TBL2
Gene Accession No. Q9R099, Q9Y4P3
Gene Alias C76179; DKFZP43N024; MGC134739; TBL2; transducin (beta)-like 2; transducin beta like 2; transducin beta-like protein 2; transducin -like 2; UNQ563/PRO1125; WBSCR13; Williams-Beuren syndrome chromosomal region 13 protein; Williams-Beuren syndrome chromosome region 13; WS beta-transducin repeats protein; WS-betaTRP; WS-bTRP
Gene Symbols TBL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TBL2(Accession Q9Y4P3), corresponding to amino acid residues L388-T438.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26608, 27368, 686610
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.