Learn More
Invitrogen™ TBL2 Polyclonal Antibody

Description
Positive test controls include: LO2, 293T, HeLa, Mouse brain, Rat pancreas. Immunogen sequence: SNGKYLATCA DDRTIRIWST KDFLQREHRS MRANVELDHA TLVRFSPDCR AFIVWLANGD TLRVFKMTKR EDGGYTFTAT PEDFPKKHKA PVIDIGIANT GKFIMTASSD TTVLIWSLKG QVLSTINTNQ MNNTHAAVSP CGRFVASCGF TPDVKVWEVC FGKKGEFQEV VRAFELKGHS AAVHSFAFSN DSRRMASVSK DGTWKLWDTD VEYKKKQDPY LLKTGRFEEA AGAAPCRLAL SPNAQVLALA SGSSIHLYNT RRGEKEECFE RVHGECIANL SFDITGRFLA SCGDRAVRLF HNTPGHRAMV EEMQGHLKRA SNESTRQRLQ Q.
Specifications
Specifications
| Antigen | TBL2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.65 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | TBL2 |
| Gene Accession No. | Q9R099, Q9Y4P3 |
| Gene Alias | C76179; DKFZP43N024; MGC134739; TBL2; transducin (beta)-like 2; transducin beta like 2; transducin beta-like protein 2; transducin -like 2; UNQ563/PRO1125; WBSCR13; Williams-Beuren syndrome chromosomal region 13 protein; Williams-Beuren syndrome chromosome region 13; WS beta-transducin repeats protein; WS-betaTRP; WS-bTRP |
| Gene Symbols | TBL2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.