Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560024 Supplier Invitrogen™ Supplier No. PA560024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LWKDVTEAEQ AEEQARQEEQ VVRQQELDNL LHEKRYLRAL GLAISLDRPH TVLTVIQAIR RDPEACEKLE ATMLRLRRDQ KEALLRFCVT WNTNSRHCHE AQAVLGVLL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

WNK is a serine/threonine kinase that plays an important role in the regulation of electrolyte homeostasis, cell signaling, survival, and proliferation. Also, acts as an activator and inhibitor of sodium-coupled chloride cotransporters and potassium-coupled chloride cotransporters respectively. Activates SCNN1A, SCNN1B, SCNN1D, and SGK1. WNK localizes to a segment of kidney involved in salt, potassium, and pH homeostasis, the distal nephron. Mutations in the WNK gene result in Pseudohypaldosteronism and Neuropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBL3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBL3
Gene Accession No. Q12788
Gene Alias 9430070M15Rik; SAZD; Tbl3; transducin (beta)-like 3; transducin beta like 3; transducin beta-like 3; transducin beta-like protein 3; UTP13; WD repeat-containing protein SAZD; WD-repeat protein SAZD
Gene Symbols TBL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBL3. Recombinant protein control fragment (Product #RP-98792).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10607
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.