Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBLR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589235 Supplier Invitrogen™ Supplier No. PA589235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QQGSAKNGEN TANGEENGAH TIANNHTDMM EVDGDVEIPP NKAVVLRGHE S Positive Samples: Jurkat, HepG2, NCI-H460, THP-1, Mouse brain, Mouse spleen, Mouse thymus; Cellular Location: Nucleus.

Chromatin organization plays a fundamental role in regulating transcription in eukaryotic cells. The nuclear receptor corepressor (N-CoR) and silencing mediator of retinoid and thyroid hormone receptors (SMRT) play a role in diverse transcriptional repression pathways. N-CoR and SMRT each exist in large protein complexes, and are thought to mediate repression, at least in part, by their ability to recruit histone deacetylases to generate repressive chromatin. HDAC3 is the main HDAC that associates with N-CoR and SMRT complexes, and its presence is thought to be essential for repression. TBLR1 is a WD-40 repeat protein that also associates with N-CoR and SMRT complexes. Although TBLR1 is thought to be a core component of both the N-CoR and SMRT complexes, its role in these complexes remains to be elucidated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBLR1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene TBL1XR1
Gene Accession No. Q8BHJ5, Q9BZK7
Gene Alias 8030499H02Rik; A630076E03Rik; AW539987; C21; C230089I12Rik; DC42; F-box-like/WD repeat protein TBL1XR1; F-box-like/WD repeat-containing protein TBL1XR1; FLJ12894; Ira1; MRD41; nuclear receptor co-repressor/HDAC3 complex subunit; nuclear receptor corepressor/HDAC3 complex subunit TBLR1; RGD1560053; TBL1-related protein 1; TBL1XR1; TBLR1; transducin (beta)-like 1 X-linked receptor 1; transducin (beta)-like 1X-linked receptor 1; transducin beta-like 1X-related protein 1
Gene Symbols TBL1XR1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-170 of human TBL1XR1 (NP_0789412).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79718, 81004
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.