Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBP Monoclonal Antibody (1TBP18)
GREENER_CHOICE

Catalog No. MA121516
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA121516 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA121516 Supplier Invitrogen™ Supplier No. MA121516
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

This antibody does not cross-react with Drosophila melanogaster, S. cerevisiae, silk worm, or Xenopus laevis. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBP
Applications Gel Shift, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1TBP18
Concentration 0.70 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Tbp
Gene Accession No. P20226, P29037
Gene Alias Gtf2d; GTF2D1; HDL4; MGC117320; MGC126054; MGC126055; RP1-191N21.3; SCA17; TATA binding protein; TA-TA binding protein 1; TATA box binding protein; TATA sequence-binding protein; TATA-binding factor; TATA-binding protein; TATA-box binding protein; TATA-box binding protein N-terminal domain; TATA-box factor; tata-box-binding; TATA-box-binding protein; TBP; TBP0; TBP1; TF2D; TFIID; TFIID core protein; transcription initiation factor TFIID TBP subunit; Unknown (protein for MGC:138100)
Gene Symbols Tbp
Host Species Mouse
Immunogen Recombinant human TBP.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117526, 21374, 6908
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.