Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144675 Supplier Invitrogen™ Supplier No. PA5144675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Hela cell lysate, HCT116 cell lysate, 293T cell lysate, HepG2 cell lysate, MCF-7 cell lysate, Jurkat cell lysate, C2C12 cell lysate, NIH/3T3 cell lysate, RAW264.7 cell lysate, mouse testis tissue lysate, rat testis tissue lysate, human bladder carcinoma tissue, human testis tissue, human colon tissue, mouse stomach tissue. Predicted band size: 38 kDa Subcellular Location: Nucleus.

RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Tbp
Gene Accession No. P20226, P29037
Gene Alias Gtf2d; GTF2D1; HDL4; MGC117320; MGC126054; MGC126055; RP1-191N21.3; SCA17; TATA binding protein; TA-TA binding protein 1; TATA box binding protein; TATA sequence-binding protein; TATA-binding factor; TATA-binding protein; TATA-box binding protein; TATA-box binding protein N-terminal domain; TATA-box factor; tata-box-binding; TATA-box-binding protein; TBP; TBP0; TBP1; TF2D; TFIID; TFIID core protein; transcription initiation factor TFIID TBP subunit; Unknown (protein for MGC:138100)
Gene Symbols Tbp
Host Species Rabbit
Immunogen Recombinant protein within human TBP aa 140-339/339.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117526, 21374, 6908
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.