Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580098
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: MCF-7 Cell, MM231 Cell, MM231 Cell.
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.
Specifications
TBP | |
Polyclonal | |
Unconjugated | |
Tbp | |
Gtf2d; GTF2D1; HDL4; MGC117320; MGC126054; MGC126055; RP1-191N21.3; SCA17; TATA binding protein; TA-TA binding protein 1; TATA box binding protein; TATA sequence-binding protein; TATA-binding factor; TATA-binding protein; TATA-box binding protein; TATA-box binding protein N-terminal domain; TATA-box factor; tata-box-binding; TATA-box-binding protein; TBP; TBP0; TBP1; TF2D; TFIID; TFIID core protein; transcription initiation factor TFIID TBP subunit; Unknown (protein for MGC:138100) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117526, 21374, 6908 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
P20226, P29037 | |
Tbp | |
A synthetic peptide corresponding to a sequence in the middle region of human TBP (227-246aa EEQSRLAARKYARVVQKLGF). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction