Learn More
TBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 220061AP150UL

Description
This antibody recognize human TBP with MW 40 kDa and mouse/rat Tbp with MW 34 kDa.
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.Specifications
TBP | |
Polyclonal | |
Unconjugated | |
TBP | |
GTF2D, GTF2D1, SCA17, TATA binding factor, TATA box binding protein, TATA box factor, TATA sequence binding protein, TBP, TF2D, TFIID | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21374, 6908 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20226, P29037 | |
TBP | |
TBP Fusion Protein Ag12383 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.