Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tbp7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Tbp7 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Tbp7 Polyclonal specifically detects Tbp7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Tbp7 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
MB67 interacting protein, MGC13687,26S protease regulatory subunit 6B, MGC23214, MGC8570, protease 26S subunit 6, proteasome (prosome, macropain) 26S subunit, ATPase, 4,26S proteasome AAA-ATPase subunit RPT3, Proteasome 26S subunit ATPase 4, S6Tat-binding protein 7, TBP-7MB67-interacting protein, TBP7MIP224 | |
PSMC4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Neuroscience, Ubiquitin Proteasome Pathway | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
5704 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LTSDQKPDVMYADIGGMDIQKQEVREAVELPLTHFELYKQIGIDPPRGVLMYGPPGCGKTMLAKAVAHHTTAAFIRVVGSEFVQKYLGEGPRMVRDVFRLAKENAPAIIFIDEIDAIATKRFDAQTGADREVQRILLEL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title