Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tbp7 Antibody (SR1808), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322626100UL
Description
Tbp7 Monoclonal antibody specifically detects Tbp7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, SerumSpecifications
Tbp7 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
SR1808 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence, Serum 1:50-1:200 | |
MB67 interacting protein, MGC13687,26S protease regulatory subunit 6B, MGC23214, MGC8570, protease 26S subunit 6, proteasome (prosome, macropain) 26S subunit, ATPase, 4,26S proteasome AAA-ATPase subunit RPT3, Proteasome 26S subunit ATPase 4, S6Tat-binding protein 7, TBP-7MB67-interacting protein, TBP7MIP224 | |
A synthesized peptide derived from human Tbp7 (Uniprot #: P43686) | |
100 μg | |
Neuroscience, Ubiquitin Proteasome Pathway | |
5704 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction