Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBPL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541110
Description
Peptide sequence: LQKLGFQVIF TDFKVVNVLA VCNMPFEIRL PEFTKNNRPH ASYEPELHPA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a protein that serves the same function as TBP and substitutes for TBP at some promoters that are not recognized by TFIID. It is essential for spermiogenesis and believed to be important in expression of developmentally regulated genes.
Specifications
| TBPL1 | |
| Polyclonal | |
| Unconjugated | |
| TBPL1 | |
| 21 kDa TBP-like protein; 21-kDA TBP-like protein; 4732475G08; AW011832; AW491032; D18347; MGC:8389; MGC:9620; second TBP of unique DNA protein; STD; STUD; TATA box binding protein-like 1; TATA box binding protein-like protein; TATA box-binding protein related factor 2; TATA box-binding protein-like 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1-like protein; TATA box-binding protein-related factor 2; TATA-box binding protein like 1; tbpl1; TBP-like 1; TBP-like factor; TBP-like protein; TBP-like protein 1; TBP-related factor 2; TBP-related protein; Tlf; TLP; Tlp21; trf; trf2; TRP; Unknown (protein for MGC:134257); zgc:92340 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 9519 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P62380 | |
| TBPL1 | |
| Synthetic peptide directed towards the middle region of human TBPL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction