Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111681 Supplier Invitrogen™ Supplier No. PA5111681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CNMPFEIRLP EFTKNNRPHA SYEPELHPAV CYRIKSLRAT LQIFSTGSIT VTGPNVKAVA TAVEQIYPFV FESRKEI Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a protein that serves the same function as TBP and substitutes for TBP at some promoters that are not recognized by TFIID. It is essential for spermiogenesis and believed to be important in expression of developmentally regulated genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBPL1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBPL1
Gene Accession No. P62380
Gene Alias 21 kDa TBP-like protein; 21-kDA TBP-like protein; 4732475G08; AW011832; AW491032; D18347; MGC:8389; MGC:9620; second TBP of unique DNA protein; STD; STUD; TATA box binding protein-like 1; TATA box binding protein-like protein; TATA box-binding protein related factor 2; TATA box-binding protein-like 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1-like protein; TATA box-binding protein-related factor 2; TATA-box binding protein like 1; tbpl1; TBP-like 1; TBP-like factor; TBP-like protein; TBP-like protein 1; TBP-related factor 2; TBP-related protein; Tlf; TLP; Tlp21; trf; trf2; TRP; Unknown (protein for MGC:134257); zgc:92340
Gene Symbols TBPL1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TBPL1. Recombinant protein control fragment (Product #RP-107837).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9519
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.