Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBR-2 Rabbit anti-Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN168725UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1687-25UG 25 μg
ABN1687-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN1687-25UG Supplier MilliporeSigma Supplier No. ABN168725UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-TBR-2, Cat. No. ABN1687, is a highly specific rabbit polyclonal antibody that targets Eomesodermin homolog (Tbr2) and has been tested in Immunohistochemistry (Paraffin), Immunofluorescence, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBR-2
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide.
Gene Alias Eomesodermin homolog;T-box brain protein 2;T-brain-2
Gene Symbols Eomes;Tbr2
Host Species Rabbit
Immunogen Conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of murine TBR-2.
Molecular Weight of Antigen ∽63 kDa observed; 74.80 kDa calculated.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_034266
Test Specificity This rabbit polyclonal antibody detects T-box brain protein 2 (TBR-2) in murine species. It targets an epitope with in 20 amino acids from the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.