Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530971 Supplier Invitrogen™ Supplier No. PA530971
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Neuro2A, GL261. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of a conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similar protein has been disrupted in mice and shown to be critical for early cortical development, and causes loss of projection neurons in the olfactory bulbs and olfactory cortex. The C-terminal region this similar protein was found to be necessary and sufficient for association with the guanylate kinase domain of calcium/calmodulin-dependent serine protein kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBR1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene TBR1
Gene Accession No. Q16650, Q64336
Gene Alias LOW QUALITY PROTEIN: T-box brain protein 1; T-box brain gene 1; T-box brain protein 1; T-box brain transcription factor 1; T-box, brain 1; T-box, brain, 1; T-box, brain, 1-like; TBR1; TBR-1; T-brain-1; TES-56
Gene Symbols TBR1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TBR1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10716, 21375, 680427
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.