Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBRG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565962
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565962 Supplier Invitrogen™ Supplier No. PA565962
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YQWVKFDVCKP GDGQLPEGLP ENDAAMSFEA FQRQIFDEDQ NDPLLPGSLD LPELQPAAFV SSYQPMYLTH EPLVDTHLQH LKSPSQGSPI Highest antigen sequence identity to the following orthologs - mouse 77%, rat 77%.

TBRG1 acts as a growth inhibitor. It can activate TP53/p53, cause G1 arrest and collaborate with CDKN2A to restrict proliferation, but it does not require either protein to inhibit DNA synthesis. TBRG1 redistributes CDKN2A into the nucleoplasm, and is involved in maintaining chromosomal stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBRG1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBRG1
Gene Accession No. Q3YBR2
Gene Alias AA408552; AA409675; mFLJ00213; Niam; Nuclear interactor of ARF and Mdm2; Tb-5; Tbrg1; transforming growth factor beta regulated gene 1; transforming growth factor beta regulator 1
Gene Symbols TBRG1
Host Species Rabbit
Immunogen Recombinant Human TBRG1. Recombinant protein control fragment (Product #RP-106652).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.