Learn More
TBRG4/CPR2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301392A
Description
The recommended shelf life for this product is 1 year from date of receipt.
May play a role in cell cycle progression.Specifications
| TBRG4/CPR2 | |
| Polyclonal | |
| Unconjugated | |
| TBRG4 | |
| CPR2, FASTKD4, FAST kinase domain-containing protein 4, FAST kinase domains 4, H_TD2522F11.8, cell cycle progression 2 protein, cell cycle progression protein 2, cell cycle progression restoration protein 2, protein TBRG4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9238 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q969Z0 | |
| TBRG4 | |
| Between 25 and 75 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.