Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBRG4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988129
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8129 Supplier Bioss Supplier No. BS7924R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBRG4 (transforming growth factor beta regulator 4), also known as CPR2 (cell cycle progression restoration protein 2) or FAST 4 (FAST kinase domain-containing protein 4), is a 631 amino acid protein that contains one RAP domain and belongs to the FAST kinase family. TBRG4 is ubiquitously expressed and may have a role in cell cycle progression. Existing as two alternatively spliced isoforms, the gene encoding TBRG4 maps to human chromosome 7p13.

Specifications

Antigen TBRG4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TBRG4
Gene Accession No. Q969Z0
Gene Alias 2310042P22Rik; AA120735; AA408001; AI527316; cell cycle progression 2 protein; Cell cycle progression protein 2; Cell cycle progression restoration protein 2; CPR2; FAST 4; FAST kinase domain-containing protein 4; FAST kinase domains 4; FASTKD4; H_TD2522F11.8; Kiaa0948; mKIAA0948; Protein TBRG4; R74877; Tb-12; TBRG4; transforming growth factor beta regulated gene 4; Transforming growth factor beta regulator 4
Gene Symbols TBRG4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TBRG4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9238
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.