Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

General Data Healthcare™ Flotation Work Station Bath
SDP

General Data's baths provide optimum specimen viewing and the dependability and safety of up-to-date temperature controlled electronics...making tissue preparation easier, safer and more efficient.

$4388.00 - $5562.00

Specifications

Controller Type Microprocessor
Certifications/Compliance UL, CSA, CE
Display LED
Dimensions (L x W x H) 18.5 x 14 x 5.4 in. (47 x 36 x 14 cm)
Length (English) 18.5 in.
View More Specs

Products 4
Catalog Number Mfr. No. Amperage Type Model Price Quantity & Availability  
Catalog Number Mfr. No. Amperage Type Model Price Quantity & Availability  
22-047-230
SDP
View Documents
General Data Healthcare™
FWSL
5 A Flotation Work Station FWS-L
Each for $5,562.00
Only null left
Add to Cart
 
22-047-234
SDP
View Documents
General Data Healthcare™
TFBL
3.5 A Tissue Flotation Bath TFB-L
Each for $4,673.00
Only null left
Add to Cart
 
22-047-232
SDP
View Documents
General Data Healthcare™
FWSS
2.5 A Flotation Work Station FWS-S
Each for $5,220.00
Only null left
Add to Cart
 
22-047-236
SDP
View Documents
General Data Healthcare™
TFBS
1.75 A Tissue Flotation Bath TFB-S
Each for $4,388.00
Only null left
Add to Cart
 
Description

Description

Ergonomically designed to provide reliability with advanced digital microprocessor control over temperature protection for preparation of paraffin embedded tissue sections

Combined with General Data’s histology consumable and reagents, these baths ensure whole ribbon sections are properly collected without cracking or over-expansion

Basic Bath

  • Bath control panel features a keypad and two LED displays
  • Arrow keys let you set and display water temperature from ambient up to 65°C (149°F)
  • Microprocessor-controlled components heat bath quickly and evenly (ambient to 65°C [149°F])
  • Second LED display switches between actual water temperature and time of day
  • Magnetic, noncontact switch functions flawlessly for years
Workstation
  • Includes everything in the basic bath, and also integrates a HISTO/Orientator™ heating block and slide dryer
  • The 2 × 2 in. (5 × 5cm) heating block can be used after removing slide from the bath to eliminate any remaining bubbles in the tissue
  • The 2 × 8 in. (5 × 20cm) slide dryer can dry several slides simultaneously
  • Both heating block and slide dryer located above control panel so as not to interfere with bath

 

Order Info

Accessories are available (See Cat. Nos. 22-047-239, -241, -242 and 22-044-630).

Specifications

Specifications

Microprocessor
LED
18.5 in.
14 in.
5.4 in.
11 lb.
100/120 V 50/60 Hz
50/60 Hz
UL, CSA, CE
18.5 x 14 x 5.4 in. (47 x 36 x 14 cm)
47 cm
36 cm
14 cm
6 kg
100/120 V
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.