Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
General Data Healthcare™ SHUR/Mark<™ Marking Pens
Shop All Triangle Biomedical Sciences Inc. (TBS) Products

Click to view available options
Color:
Red
Description
- For cassettes and slides
- Fast drying
- Simply depress tip until saturated
- Will not smudge or smear
- Reagent resistant to histology solvents, especially alcohol and xylene
- Replaceable tip eliminates clogged or worn out tips
Specifications
Specifications
Product Type | Marking Pen |
Color | Red |
For Use With (Application) | For labeling slides or cassettes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction