Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

General Data Healthcare™ SHUR/Stain™ Manual Stainers
SDP

Catalog No. p-4374821
Click to view available options
Positions:
3
6
12
Width (Metric):
20.3 cm
37.5 cm
71.8 cm
Length (Metric):
15.6 cm
17.8 cm

Manual stain rack with individual lids for use during extended inactivity

  • Metal extruded frames inter-lock for back-to-back connection of multiple racks
  • See-through lexan cover included
  • Plastic dishes are solvent resistant and include lids for limiting evaporation
  • With digital timer and label strips to record reagent expiration dates

SS-3L:

  • 3 positions
  • Includes two white 250mL wells (SS-WLW), one green solvent-resistant 250mL well (SS-WLG), one 24-position plastic slide carrier (SS-SRG-24)
  • See-through Lexan™ cover
  • Three 250mL wells for 1 x 3 in. slides
  • D x W x H: 6 x 8 x 7 in. (160 x 203 x 178mm)

SS-6L:

  • 6 positions
  • Includes four white 250mL wells (SS-WLW), two green solvent-resistant 250mL well (SS-WLG), one 24-poisition plastic slide carrier (SS-SRG-24)
  • See-through plastic lid
  • Six 250mL wells for 1 x 3 in. slides
  • D x W x H: 6 x 15 x 6 in. (160 x 375 x 156mm)

SS-12L:

  • 12 positions
  • Includes eight white 250mL wells (SS-WLW), four green solvent-resistant 250mL well (SS-WLG), one 24-poisition plastic slide carrier (SS-SRG-24)
  • See-through plastic lid
  • Twelve 250mL wells for 1 x 3 in. slides
  • D x W x H: 6 x 28 x 6 in. (160 x 718 x 156mm)

Specifications

Includes Eight white 250mL wells (SS-WLW), four green solvent resistant 250mL well (SS-WLG), one 24-poisition plastic slide carrier (SS-SRG-24), See-through lexan cover
Material Plastic
Positions 12
Dimensions (L x W) 6 x 28 in. (15.6 x 71.8cm)
Length (Metric) 15.6 cm
Length (English) 6 in.
Width (Metric) 71.8 cm
Width (English) 28 in.
Depth (English) 6 in.
Depth (Metric) 16 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.