Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBX1 Antibody (OTI1C2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274470AF700
Description
TBX1 Monoclonal specifically detects TBX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TBX1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TBX1 | |
Full length recombinant protein of human TBX1 (NP_542377) produced in HEK293T cells. | |
0.1 mL | |
6899.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1C2 | |
Western Blot, Immunohistochemistry | |
brachyury, CAFS, CTHM, DGCR, DGS, DORV, T-box 1, T-box 1 transcription factor C, T-box protein 1, T-box transcription factor TBX1, TBX1C, Testis-specific T-box protein, TGA, VCFS | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction