Learn More
TBX1 Mouse anti-Human, Clone: OTI1C2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF507326

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TBX1 is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product shares 98% amino acid sequence identity with the mouse ortholog. DiGeorge syndrome (DGS)/velocardiofacial syndrome (VCFS), a common congenital disorder characterized by neural-crest-related developmental defects, has been associated with deletions of chromosome 22q11.2, where this gene has been mapped. Studies using mouse models of DiGeorge syndrome suggest a major role for this gene in the molecular etiology of DGS/VCFS. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
TBX1 | |
Monoclonal | |
Unconjugated | |
TBX1 | |
CAFS, CATCH22, CTHM, DGCR, DGS, DORV, TBX1C, TGA, VCF, VCFS | |
Mouse | |
Affinity Chromatography | |
RUO | |
6899 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43435 | |
TBX1 | |
Full length human recombit protein of human TBX1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.