Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TBX1, Mouse, Unlabeled, Clone: 7E12.1,

Catalog No. MABN720MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN720MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN720MI Supplier MilliporeSigma™ Supplier No. MABN720
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

T-box transcription factor TBX1 (UniProt O43435; also known as T-box protein 1, Testis-specific T-box protein) is encoded by the TBX1 (also known as CTHM, DGS, TBX1C, VCFS) gene (Gene ID 6899) in human. TBX1 blongs to the T-box family of transcription factors that play important roles in the formation of tissues and organs during embryonic development. TBX1 is required for the second heart field (SHF) development for outflow tract development by regulating proliferation and differentiation of SHF cells. TBX1 is also shown to negatively regulate thymic epithelial cell (TEC) growth and differentiation in mice, extinction of Tbx1 gene expression in the third pharyngeal pouch (3rd pp) endoderm is a prerequisite for thymus organogenesis. Human TBX mutations cause DiGeorge syndrome, characterized by parathyroid hypoplasia, thymic hypoplasia, defects in the outflow tract of the heart, and craniofacial anomalies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX1
Applications Western Blot
Classification Monoclonal
Clone 7E12.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Mouse monoclonal IgG2aκ hybridoma culture supernatant without preservatives.
Gene TBX1, CTHM, DGS, TBX1C, VCFS
Gene Accession No. O43435
Host Species Mouse
Immunogen GST-tagged recombinant human TBX1 C-terminal fragment.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_542377
Test Specificity Clone 7E12.1 was raised against a recombinant fragment derived from TBX1 spliced isoform A (Uniprot O43435-1), cross-reactivity toward spliced isoform B and isoform C (TBX1C) is possible, but has not been determined.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Culture Supernatant
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.