Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526389
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526389 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526389 Supplier Invitrogen™ Supplier No. PA526389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TBX1 Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

TBX1 is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product shares 98% amino acid sequence identity with the mouse ortholog. DiGeorge syndrome (DGS)/velocardiofacial syndrome (VCFS), a common congenital disorder characterized by neural-crest-related developmental defects, has been associated with deletions of chromosome 22q11.2, where this gene has been mapped. Studies using mouse models of DiGeorge syndrome suggest a major role for this gene in the molecular etiology of DGS/VCFS. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TBX1
Gene Accession No. O43435
Gene Alias brachyury; CAFS; CATCH22; CTHM; DGCR; DGS; DORV; LOW QUALITY PROTEIN: T-box transcription factor TBX1; T-box 1; T-box 1 transcription factor C; T-box protein 1; T-box transcription factor 1; T-box transcription factor TBX1; T-box transcription factor TBX1; LOW QUALITY PROTEIN: T-box transcription factor TBX1; Tbx1; TBX1C; Testis-specific T-box protein; TGA; VCF; VCFS
Gene Symbols TBX1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 327-356 amino acids from the C-terminal region of human TBX1.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.