Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575214
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
TBX1 is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product shares 98% amino acid sequence identity with the mouse ortholog. DiGeorge syndrome (DGS)/velocardiofacial syndrome (VCFS), a common congenital disorder characterized by neural-crest-related developmental defects, has been associated with deletions of chromosome 22q11.2, where this gene has been mapped. Studies using mouse models of DiGeorge syndrome suggest a major role for this gene in the molecular etiology of DGS/VCFS. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
TBX1 | |
Polyclonal | |
Unconjugated | |
TBX1 | |
brachyury; CAFS; CATCH22; CTHM; DGCR; DGS; DORV; LOW QUALITY PROTEIN: T-box transcription factor TBX1; T-box 1; T-box 1 transcription factor C; T-box protein 1; T-box transcription factor 1; T-box transcription factor TBX1; T-box transcription factor TBX1; LOW QUALITY PROTEIN: T-box transcription factor TBX1; Tbx1; TBX1C; Testis-specific T-box protein; TGA; VCF; VCFS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6899 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O43435 | |
TBX1 | |
Synthetic peptide corresponding to amino acids 321-370 of Human TBX1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction