Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBX10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP21341725UL
Description
TBX10 Polyclonal specifically detects TBX10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TBX10 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
T-box 10, T-box 7, T-box protein 10, T-box transcription factor TBX10, T-box-containing transcriptional activator, TBX13, TBX7 | |
Rabbit | |
Affinity Purified | |
RUO | |
347853 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TBX10 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: PARSHSSLSPCVLKGATDREKDPNKASASTSKTPAWLHHQLLPPPEVLLAPATYRPVTYQSLYSGAPSHLGIPRTR | |
25ul | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction