Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584106 Supplier Invitrogen™ Supplier No. PA584106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSPPTFHLAP NTFNVGCRES QLCNLNLSDY PPCARSNMAA LQSYPGLSDS GYNRLQSGTT SATQPSETFM PQRTPSLISG IPTPPSLPGN SKMEAYGGQL GSFPTSQFQY VMQ Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.

TBX15 is a probable transcriptional regulator involved in developmental processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX15
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBX15
Gene Accession No. Q96SF7
Gene Alias T-box 14; T-box 15; T-box protein 14; T-box protein 15; T-box transcription factor TBX14; T-box transcription factor TBX15; TBX14; TBX15
Gene Symbols TBX15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TBX15. Recombinant protein control fragment (Product #RP-89307).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.