Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBX15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113201
Description
TBX15 Polyclonal Antibody for Western Blot, ELISA
TBX15 is a probable transcriptional regulator involved in developmental processes.
Specifications
TBX15 | |
Polyclonal | |
Unconjugated | |
TBX15 | |
T-box 14; T-box 15; T-box protein 14; T-box protein 15; T-box transcription factor TBX14; T-box transcription factor TBX15; TBX14; TBX15 | |
Rabbit | |
Protein G | |
RUO | |
6913 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96SF7 | |
TBX15 | |
Recombinant Human T-box transcription factor TBX15 protein (1-496AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction