Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX15/TBX18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536143 Supplier Invitrogen™ Supplier No. PA536143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 64 kDa. Purity is >95% by SDS-PAGE.

T-box transcription factors are a group of phylogenetically conserved genes that contain a uniquely defining DNA-binding domain, the T-box domain. These genes are believed to be involved in the regulation of development processes. In mouse embryo development, Tbx15 is expressed at the beginning of day 9.5, primarily in the craniofacial region and in the developing limbs. Twelve human T-box genes have been isolated so far. TBX 18 has been reported to be involved in numerous development processes and to act as an antiapoptotic factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX15/TBX18
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TBX18
Gene Accession No. O70306, O95935, Q96SF7, Q9EPZ6
Gene Alias 2810012F10Rik; 2810404D13Rik; CAKUT2; de; droopy ear; mmTBx8; T-box 14; T-box 15; T-box 18; T-box protein 14; T-box protein 15; T-box protein 18; T-box transcription factor TBX14; T-box transcription factor TBX15; T-box transcription factor TBX18; T-box18; TBX14; TBX15; Tbx18; Tbx8
Gene Symbols TBX15, TBX18
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 131-180 of human TBX15.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21384, 295315, 315870, 6913, 76365, 9096
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.