Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX15/TBX18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103258 Supplier Invitrogen™ Supplier No. PA5103258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TBX15/18.

T-box transcription factors are a group of phylogenetically conserved genes that contain a uniquely defining DNA-binding domain, the T-box domain. These genes are believed to be involved in the regulation of development processes. In mouse embryo development, Tbx15 is expressed at the beginning of day 9.5, primarily in the craniofacial region and in the developing limbs. Twelve human T-box genes have been isolated so far. TBX 18 has been reported to be involved in numerous development processes and to act as an antiapoptotic factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX15/TBX18
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TBX18
Gene Accession No. O70306, O95935, Q96SF7, Q9EPZ6
Gene Alias 2810012F10Rik; 2810404D13Rik; CAKUT2; de; droopy ear; mmTBx8; T-box 14; T-box 15; T-box 18; T-box protein 14; T-box protein 15; T-box protein 18; T-box transcription factor TBX14; T-box transcription factor TBX15; T-box transcription factor TBX18; T-box18; TBX14; TBX15; Tbx18; Tbx8
Gene Symbols TBX15, TBX18
Host Species Rabbit
Immunogen A synthesized peptide derived from human TBX18(Accession O95935), corresponding to amino acid residues P170-P220.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21384, 295315, 315870, 6913, 76365, 9096
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.