Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBX18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569067
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 79%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
The T-box (Tbx) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The Tbx gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. Tbx genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in Tbx genes are associated with the onset of several human diseases. Our understanding of functional mechanisms of Tbx products has come mainly from the prototypical T/Brachyury, which is a transcription activator. The Tbx genes constitute a family of transcriptional regulatory genes that are implicated in a variety of developmental processes ranging from the formation of germ layers to the organizational patterning of the central nervous system.
Specifications
| TBX18 | |
| Polyclonal | |
| Unconjugated | |
| TBX18 | |
| 2810012F10Rik; 2810404D13Rik; CAKUT2; LOW QUALITY PROTEIN: T-box transcription factor TBX18; T-box 18; T-box protein 14; T-box protein 15; T-box protein 18; T-box transcription factor 18; T-box transcription factor TBX18; T-box18; TBX14; TBX15; Tbx18 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 315870 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| TBX18 | |
| synthetic peptide directed towards the N-terminal region of rat TBX18. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction