Learn More
TBX19 (Tpit) Mouse anti-Human, Clone: OTI1C9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800675

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Mutations in this gene were found in patients with isolated deficiency of pituitary POMC-derived ACTH, suggesting an essential role for this gene in differentiation of the pituitary POMC lineage. ACTH deficiency is characterized by adrenal insufficiency symptoms such as weight loss, lack of appetite (anorexia), weakness, nausea, vomiting, and low blood pressure.Specifications
TBX19 (Tpit) | |
Monoclonal | |
Unconjugated | |
TBX19 | |
TBS19, TPIT, dJ747L4.1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9095 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1C9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O60806 | |
TBX19 | |
Human recombit protein fragment corresponding to amino acids 1-238 of human TBX19 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.