Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX2/3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116010 Supplier Invitrogen™ Supplier No. PA5116010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TBX 2/3.

TBX2 (T-Box Transcription Factor 2) is a Protein Coding gene. Diseases associated with TBX2 include Vertebral Anomalies And Variable Endocrine And T-Cell Dysfunction and Ulnar-Mammary Syndrome. Among its related pathways are Heart Development and Inhibition of Ribosome Biogenesis by p14(ARF). Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity and RNA polymerase II proximal promoter sequence-specific DNA binding. An important paralog of this gene is TBX3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX2/3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TBX3
Gene Accession No. O15119, P70324, Q13207, Q60707, Q7TST9
Gene Alias bladder cancer related protein XHL; D5Ertd189e; T-box 2; T-box 3; T-box 3 (ulnar mammary syndrome); T-box protein 2; T-box protein 3; T-box transcription factor TBX2; T-box transcription factor TBX3; TBX2; TBX3; TBX3/2; TBX3-ISO; UMS; XHL
Gene Symbols Tbx2, TBX3
Host Species Rabbit
Immunogen A synthesized peptide derived from human TBX3(Accession O15119), corresponding to amino acid residues A283-N333.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21385, 21386, 303398, 353305, 6909, 6926
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.