Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587747
Description
Positive Samples: MCF7; Cellular Location: Nucleus Immunogen sequence: GSLSRSPFLG SARPRLRFSP YQIPVTIPPS TSLLTTGLAS EGSKAAGGNS REPSPLPELA LRKVGAPSRG ALSPSGSAKE AANELQSIQR LVSGLESQRA LSPGRESPK.
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product is the human homolog of mouse Tbx2, and shares strong sequence similarity with Drosophila omb protein. Expression studies indicate that this gene may have a potential role in tumorigenesis as an immortalizing agent. Transcript heterogeneity due to alternative polyadenylation has been noted for this gene.
Specifications
TBX2 | |
Polyclonal | |
Unconjugated | |
TBX2 | |
FLJ10169; LOW QUALITY PROTEIN: T-box transcription factor TBX2; T-box 2; T-box 2 protein; T-box 2 transcription factor; T-box protein 2; T-box transcription factor 2; T-box transcription factor TBX2; TBX2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
21385, 303398, 6909 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.68 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13207, Q60707 | |
TBX2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 604-712 of human TBX2 (NP_0059853). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction