Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599727
Description
Antibody detects endogenous levels of total TBX2.
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product is the human homolog of mouse Tbx2, and shares strong sequence similarity with Drosophila omb protein. Expression studies indicate that this gene may have a potential role in tumorigenesis as an immortalizing agent. Transcript heterogeneity due to alternative polyadenylation has been noted for this gene.
Specifications
| TBX2 | |
| Polyclonal | |
| Unconjugated | |
| TBX2 | |
| FLJ10169; LOW QUALITY PROTEIN: T-box transcription factor TBX2; T-box 2; T-box 2 protein; T-box 2 transcription factor; T-box protein 2; T-box transcription factor 2; T-box transcription factor TBX2; TBX2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21385, 6909 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13207, Q60707 | |
| TBX2 | |
| A synthesized peptide derived from human TBX2(Accession Q13207), corresponding to amino acid residues I630-S680. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction