Learn More
TBX2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 223461AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-3842
Only null left
Add to Cart
Description
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product is the human homolog of mouse Tbx2, and shares strong sequence similarity with Drosophila omb protein. Expression studies indicate that this gene may have a potential role in tumorigenesis as an immortalizing agent. Transcript heterogeneity due to alternative polyadenylation has been noted for this gene.
Specifications
Specifications
TBX2
Polyclonal
Unconjugated
TBX2
T box 2, T box protein 2, TBX2
Rabbit
Antigen Affinity Chromatography
RUO
21385, 6909
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13207, Q60707
TBX2
TBX2 Fusion Protein Ag17860
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.