Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113564 Supplier Invitrogen™ Supplier No. PA5113564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RANAFSIAAL MSSGGSKEKE ATENTIKPLE QFVEKSSCAQ PLGELTSLDA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.

TBX20 is a member of the T-box transcription factor family expressed in the developing heart, eye, ventral neural tube, and limbs, indicating a possible role in regulating development of these tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX20
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TBX20
Gene Accession No. Q9UMR3
Gene Alias 9430010M06Rik; AL022859; ASD4; cb826; H15-related T-box transcription factor hrT; hrt; T-box 20; T-box protein 20; T-box transcription factor; T-box transcription factor 20; T-box transcription factor TBX20; T-box transcription factor TBX20-like; Tbx12; TBX20; whz; ZF-TBX20
Gene Symbols TBX20
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Human TBX20 (aa 14-63).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57057
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.