Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596813 Supplier Invitrogen™ Supplier No. PA596813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDGLLETYPW RPSFTLDFKT FGADTQSGSS GSSPVTSSGG APSPLNSLLS PLCFSPMFHL PTSSLGMPCP EAYLPNVNLP LCYKICPTNF WQQQPLVLPA PERLASSNSS QSLAPLMMEV PMLSSLGVTN SKSGSSEDSS DQYLQAPNST NQMLYGLQSP GNIFLPNSIT PEALSCSFHP SYDFYRYNFS MPSRLISGSN HLKVNDDSQV SFGEGKCNHV HWYPAINHYL; Positive Samples: Rat liver; Cellular Location: Nucleus.

The T-box (TBX) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The TBX gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. TBX genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in TBX genes are associated with the onset of several human diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX22
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TBX22
Gene Accession No. Q8K402, Q9Y458
Gene Alias ABERS; CLPA; CPX; D230020M15Rik; dJ795G23.1; T-box 22; T-box protein 22; T-box transcription factor TBX22; TBOX22; Tbx 22; TBX22; TBXX
Gene Symbols TBX22
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 291-520 of human TBX22.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245572, 302369, 50945
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.